missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bovine IFN-beta Protein

Product Code. 18697148 Shop All Bio Techne Products
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 18697148

Brand: Novus Biologicals™ NBP262424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA
Formulation PBS
Gene ID (Entrez) 3456
Molecular Weight (g/mol) MolecularWeight-theroretical: 19.4 kDa
Quantity 5 μg
Storage Requirements Store at -20°C.
Gene Alias Fibroblast interferon, IFBIFNB, IFF, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Gene Symbol IFNB1
Reconstitution Reconstitute with 20 μL sterile PBS containing 0.1% carrier protein. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Research Category Apoptosis
Purity or Quality Grade >95%, by SDS-PAGE
Protein IFN-beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.