missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation Protein-Pak CM 15HR Column

Waters Advanced Purification (AP) Column containing Protein-Pak CM (carboxymethyl) weak cation exchange ligand coupled to polymethacrylate particles.

Brand:  Waters Corporation WAT039787

Additional Details : Weight : 0.50000kg

Product Code. 13119189

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Waters Advanced Purification (AP) Column (5 x 50 mm) containing Protein-Pak CM (carboxymethyl) weak cation exchange ligand coupled to 15 μm polymethacrylate particles.
Specifications

Specifications

5 mm
Spherical
1000 Å
Column
50 mm
15 μm
1/Ea.
Protein
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions