Learn More
Abeba™ Protektor Line 1630 Shoes
Leather upper construction includes reflective material.
Brand:  Abeba™ 1630/37
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15637427
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Instep area with lacing
  • Cushioned ankle
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Leather
Slip-resistant
1630
White
37
Polyurethane
Steel