Learn More
Abeba™ Protektor Line 1630 Shoes
Product Code. 15687427
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15687427
Brand: Abeba™ 1630/42
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Leather upper construction includes reflective material.
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Instep area with lacing
  • Cushioned ankle
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S2 SRC
Color White
Material Leather
Size 42
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1630
Toe Material Steel