Learn More
Abeba™ Protektor Line 1635 Shoes
Leather upper construction includes reflective material.
Brand:  Abeba™ 1635/39
Additional Details : Weight : 1.33000kg
 View more versions of this product
Product Code. 15617457
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather white
  • Steel toe cap
  • Penetration resistant steel midsole
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20345:2011 S3 SRC
Leather
Slip-resistant
1635
White
39
Polyurethane
Steel