Learn More
Hamilton™ PRP-1 5 μm 100 Å 2.1 x 150 mm
PRP-1 columns have inherent reversed-phase separation qualities that eliminate the requirement for a stationary phase coating. Hydrolysis of the stationary phase coating is often the limiting factor in column lifetime, so the lack of coating on the PRP-1 resin extends the longevity of the column. PRP-1 also have exceptional sample recovery (≥ 95%).
Brand: Hamilton™ 79366
Additional Details : Weight : 0.13900kg
Description
Specification Sheet
Separation Mode | Reversed-Phase |
---|---|
Stationary Phase | PRP-1 |
Format | HPLC Column |
Hardware Inner Diameter | 2.1 mm |
Hardware Length | 150 mm |
Particle Size | 5 μm |
USP Classification | USP L21 |
Packing Material Type | PSDVB |
Pore Size | 100 Å |
Hardware Material | Stainless Steel |
Max Pressure | 5,000 psi |
Mobile Phase Limits | pH 1-13. 0-100% aqueous, organic modifier |
Buffer Strength | 0.0-0.5N |
Temp Limits | 5-85°C |
Restoration | Run a gradient of 100% water to 100% acetonitrile. Repeat three times |
Compound Classification | Organics |
UOM | 1 EACH |
Specifications
HPLC Column | |
150 mm | |
5 μm | |
100 Å | |
USP L21 | |
Reversed Phase |
2.1 mm | |
5,000 psi | |
pH 1-13. 0-100% aqueous, organic modifier | |
5° to 85°C | |
PSDVB | |
Stainless Steel |