missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ PRP-h5 5 μm 300 Å 2.1 x 150 mm
Pentafluoro reversed-phase column for unique selectivity. The PRP-h5 utilizes the PRP-3 as its base with a pentafluorinated modification, making it more hydrophobic in nature.
Brand: Hamilton™ 79271
Additional Details : Weight : 1.00000kg
Description
Specification Sheet
Separation Mode | Reversed-Phase |
---|---|
Stationary Phase | PRP-h5 |
Format | HPLC Column |
Hardware Inner Diameter | 2.1 mm |
Hardware Length | 150 mm |
Particle Size | 5 μm |
Packing Material Type | PSDVB |
Pore Size | 300 Å |
Hardware Material | Stainless Steel |
Max Pressure | 5,000 psi |
Mobile Phase Limits | pH 1-13. 0-100% aqueous, organic modifier |
Buffer Strength | 0.0-0.5N |
Temp Limits | 5-85°C |
Restoration | Run a gradient of 100% water to 100% acetonitrile. Repeat three times |
Compound Classification | Peptides/Proteins |
UOM | 1 EACH |
Specifications
HPLC Column | |
150 mm | |
5 μm | |
300 Å | |
PSDVB | |
Stainless Steel |
2.1 mm | |
5,000 psi | |
pH 1-13. 0-100% aqueous, organic modifier | |
5° to 85°C | |
Reversed Phase |