missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSEM 89S
GREENER_CHOICE

Product Code. 18769406
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18769406

Brand: Tocris Bioscience™ 6426/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 chimeric ion channel agonist

PSEM 89S is a PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 chimeric ion channel agonist (EC50 values are 3.4 and 2.2 μM, respectively). Activates cortical neurons expressing PSAML141F,Y115F-5-HT3 chimeric ion channels and inhibits activity of neurons expressing PSAML141F-GlyR in vitro. Silences hypothalamic AGRP neurons transfected with PSAML141F-GlyR chimeric ion channels in mice in vivo, leading to suppression of feeding. Brain penetrant.

Plasmid vectors for the transfection of cells with PSAML141F-GlyR and PSAML141F,Y115F-5-HT3 are available from Addgene.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3 S)-1-Azabicyclo[2.2.2 ]oct-3-yl-2,5-dimethoxybenzamide trifluoroacetate
CAS 1336913-03-1
Quantity 25 mg
Target Pharmacologically Selective Effector Molecules/PSEM
Molecular Formula C16H22N2O3.CF3CO2H
Formula Weight 404.38
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.