Learn More
RETSCH PTFE Adapter Rack
Shop All Retsch Products
Click to view available options
:
For Use With 10 Reaction Vials 1.5 and 2mL
Specifications
Specifications
| Product Type | Adapter Rack |
| For Use With (Equipment) | 10 Reaction Vials 1.5 and 2 mL |
| Material | PTFE |