Learn More
Bochem™ PTFE Bent Tip Forceps
Click to view available options
Length (Metric):
115 mm
145 mm
Unit Size:
Each
Specifications
Specifications
| Tip Style | Bent |
| No. per Pack | 10/Pack |
| Material | PTFE |
| Length (Metric) | 115 mm |
| Product Type | Forceps |
| Includes | Guide pin |
| Type | Forceps |