Learn More
Thermo Scientific™ PTFE Bottles
Product Code. p-6698022
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1 L
2000 mL
Height (Metric):
26 mm
61 mm
76 mm
80 mm
88 mm
100 mm
115 mm
150 mm
185 mm
240 mm
Unit Size:
Each
Product Code. 15426185
Brand: Thermo Scientific™ 038109.KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with aggressive or valuable reagents
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Inert and leak-free
  • Flexible and with PTFE cap
TRUSTED_SUSTAINABILITY
Specifications
Capacity 10 mL
Capacity (Metric) 10 mL
Material PTFE
Height (Metric) 26 mm
Product Type Bottle
Diameter (Metric) 12 mm