Learn More
Bohlender™ PTFE GL Tube Fittings With GL Thread
Use these fitting as reduction for connecting different diameters of tubes or tubing. Bohlender™ PTFE GL Tube Fittings With GL Thread are ideal for distributing liquids or gases.
74.94€ - 119.78€
Specifications
Product Type | GL Tube Fittings |
---|---|
Certifications/Compliance | FDA Conform |
Material | PTFE |
For Use With (Application) | For connecting different diameters of tubes or tubing |
Min. Operating Temperature (Metric) | -200°C |
Product Code | Brand | Overall Length (Metric) | Thread Size | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Length (Metric) | Thread Size | Shape | Price | Quantity & Availability | |||
10073341
|
Bohlender™
D540-14 |
54 mm | GL 14 | T-shape, three connections |
74.94€
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
10557081
|
Bohlender™
D539-25 |
51 mm | GL 25 | Elbow-shaped, two connections |
119.78€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
10400152
|
Bohlender™
D540-18 |
56 mm | GL 18 | T-shape, three connections |
100.33€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
Description
- Tube fitting T-shaped made of PTFE
- Three connections with GL thread
- Universal chemical resistance, the product is only exposed to PTFE
- Suitable for vacuum
Specifications
GL Tube Fittings | |
PTFE | |
-200°C | |
10 bar |
FDA Conform | |
For connecting different diameters of tubes or tubing | |
1 Each | |
250°C |