Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Single-sided crosshead
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
10 mm
14 mm
17 mm
25 mm
40 mm
60 mm
Unit Size:
Each
Pack of 10
Pack of 5
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Height (Metric) | 15mm |
| Diameter (Metric) | 25 mm |
| Quantity | 2/Pk. |