Learn More
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Product Code. 11725064 Shop All SP Scienceware Products
Click to view available options
Quantity:
3 sizes
3 sizes (2 each size)
7 sizes
Shape:
Octagon
Sphere
Unit Size:
Pack of 22
Pack of 6
Product Code. 11725064
Brand: Bel-Art™ F371600000
Only null left
Add to basket
Add to basket
Supplied in a convenient storage box and provide instant availability of PTFE encapsulated Alnico magnets.
  • Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Order Info
DimensionsLxW
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2015
Color White
Dimensions (L x W x H) 25.4 x 8mm, 38.1 x 8mm, 50.8 x 8mm (Two Each)
For Use With (Equipment) Vials, Micro test tubes, Centrifuge tubes
Material Alnico Magnet, PTFE
Quantity 3 sizes (2 each size)
Shape Octagon