Learn More
Adtech Polymer Engineering™ PTFE Microtubing
Click to view available options
Diameter (Metric) Inner:
0.3 mm
0.56 mm
0.81 mm
1.06 mm
1.32 mm
Diameter (Metric) Outer:
0.76 mm
1.07 mm
1.63 mm
1.68 mm
1.93 mm
Unit Size:
30m
Each
Description
- Maximum temperature of 260°C (melting point at 330°C)
- Non-stick characteristics
- Low friction surface
- Very high resistance to chemicals and solvents
- Very high electrical resilience, good for insulation
- Physiologically neutral
- Non-polluting and oderless
- Sterilizable and autoclavable
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.81 mm |
| Material | PTFE |
| Diameter (Metric) Outer | 1.63 mm |
| Overall Length (Metric) | 30 m |
| For Use With (Application) | Hot work, transferring corrosive chemicals |
| Min. Operating Temperature (Metric) | -240°C |
| Packaging Quantity | 1 Pack |
| Color | Translucent |
| Show More |