Learn More
Wheaton™ PTFE Pestles
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
1 mL
3 mL
5 mL
15 mL
Unit Size:
Pack of 2
Specifications
Specifications
| Capacity (Metric) | 15 mL |
| Diameter (Metric) Outside | 18mm |
| Length (Metric) | 240mm |
| Length (Metric) Pestle | 240mm |
| Length (Metric) Tube | 114mm |
| Material | PTFE |