Learn More
OHAUS™ PTFE Spinbar
Shop All Ohaus Products
Click to view available options
Length (Metric):
2.5 cm
3.8 cm
7 cm
10 cm
Unit Size:
Each
Description
- Constructed of PTFE
- 10 cm spinbar includes a pivot ring
Specifications
Specifications
| Color | White |
| Description | Spinbar 10 cm with Pivot Ring PTFE |
| Includes | Pivot Ring |
| Length (Metric) | 10 cm |
| Material | PTFE |