Learn More
Bohlender™ BOLA™ PTFE Straight Tube Fitting With GL Thread
Use this fitting to connect different diameters of tubes or tubing. Bohlender™ BOLA™ PTFE Straight Tube Fittings With GL Thread feature universal chemical resistance, ideal for distributing liquids or gases.
47.97€ - 294.68€
Specifications
Product Type | Straight Tube Fitting |
---|---|
Certifications/Compliance | FDA Conform |
Material | PTFE |
For Use With (Application) | For connecting different diameters of tubes or tubing |
Min. Operating Temperature (Metric) | -200°C |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
10134581
|
Bohlender™
D538-18 |
GL 18 |
48.55€
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
10359321
|
Bohlender™
D538-32 |
GL 32 |
216.24€
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
10033331
|
Bohlender™
D538-45 |
GL 45 |
294.68€
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
10175111
|
Bohlender™
D538-14 |
GL 14 |
47.97€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
- Straight tube fitting made of PTFE
- Two connections with GL thread
- The product is only exposed to PTFE
- Temperature range: -200 to +250°C
Specifications
Straight Tube Fitting | |
PTFE | |
-200°C | |
10 bar |
FDA Conform | |
For connecting different diameters of tubes or tubing | |
1 Each | |
250°C |