Learn More
Bohlender™ PTFE Temperature Probes Lemo
Product Code. 10318851
Click to view available options
Length (Metric):
160 mm
360 mm
560 mm
Unit Size:
Each
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10318851
Brand: Bohlender™ P176020
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Material PTFE
Temperature (Metric) Range -200°C to +250°C
Type Temperature Probe
Length (Metric) 360 mm
Connector Types Lemo
Length (Metric) Cable 300 mm