Learn More
Fisherbrand™ PTFE Triangle with Rib Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
12 mm
25 mm
35 mm
50 mm
80 mm
Diameter (Metric):
6 mm
8 mm
9 mm
12 mm
18 mm
Unit Size:
Pack of 10
Pack of 5
Description
- Encapsulated PTFE
- Alnico V Magnet Core
- Polished finish to reduce pick-up and contamination
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Easy to clean
Specifications
Specifications
| Description | Fisherbrand PTFE Triangle with Rib Stir Bar |
| For Use With (Equipment) | Uneven surfaces |
| Temperature Range (Metric) | -200°C to +280°C |
| Diameter (Metric) | 9 mm |
| Length (Metric) | 35 mm |
| Material | PTFE with Alnico V Magnet Core |
| Quantity | 1 |