Learn More
Sino Biological™ PTH Recombinant Human Protein, N-GST Tag
Expressed from DNA sequence encoding human PTH (Ser 32-Phe 65) fused to GST tag at N-terminus
Marque: Sino Biological™ 13192H09E50
Informations supplémentaires : Poids : 0.01000kg
Description
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Spécification
Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications | |
5741 | |
≥90% by SDS-PAGE | |
Human | |
<1 EU/μg | |
PTH | |
Hormones and Receptors | |
PTH Recombinant Human Protein |
Lyophilized | |
31kd | |
50μg | |
Truncated, Recombinant | |
Parathyroid Hormone | |
Assay Standard | |
E.coli | |
GST Tag |
For Research Use Only. Not for use in diagnostic procedures.