Learn More
Moldex™ Spark Plugs™ MoldexStation Ear Plug Dispenser System
Product Code. p-8009805
Click to view available options
Size:
Large
Includes:
500 pairs Moldex™ PU-foam earplugs
Unit Size:
500 pairs
Product Code. 11915537
Brand: Moldex™ 785001
Only null left
Add to basket
Add to basket
Deliver plugs without the mess of individual paper/polybag packaging. Moldex™ Spark Plugs™ MoldexStation includes earplugs made of soft, comfortable material which when rolled becomes firmer for easier insertion.
  • Includes Spark Plugs™ tapered design disposable earplugs
  • Plugs remain hygienic as the dispenser mechanism is automatically replaced with every refill
  • Available with all Moldex PU-foam disposable earplugs
  • Available with 250 and 500 pairs of plugs
  • The mounting bracket comes separately and is needed for all Moldex Stations
  • Earplugs are tested and certified to EN 352-2:2002
Specifications
Size Large
Quantity 500/Pk.
Product Type Ear Plug Dispenser
Mounting Mounting bracket comes separately; required for all MoldexStations
Includes 500 pairs Moldex™ PU-foam earplugs