Learn More
RETSCH Pure Titanium Ring Sieve For Ultra Centrifugal Mill ZM 200
Product Code. 10436943 Shop All Retsch Products
Click to view available options
Pore Size:
80 μm
1 mm
120 μm
1.5 mm
2 mm
200 μm
250 μm
4 mm
500 μm
750 μm
Unit Size:
Each
Product Code. 10436943
Brand: RETSCH 03.647.0277
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Quantity 1 Ea.
Pore Size 250 μm
Type Ring Sieve
Material Titanium
For Use With (Equipment) Ultra Centrifugal Mill ZM 200