Learn More
Invitrogen™ PureLink™ Pro 96 total RNA Purification Kit
Product Code. 10604833
Click to view available options
Quantity:
2 x 1 L
384 Preps
5 x 1 L
750 mL
Product Type:
RNA Lysis Buffer
RNA Purification Kit
RNA Wash Buffer I
RNA Wash Buffer II (5X)
Unit Size:
750mL
Pack of 2
Pack of 4
Pack of 5
Product Code. 10604833
Brand: Invitrogen™ 12173032
The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes.
The PureLink Pro 96 total RNA Purification Kit provides high-throughput isolation of high-quality total RNA from the widest range of sample types and sizes. Isolate high yields of total RNA with low genomic DNA contamination from bacteria, yeast, plant, or mammalian samples.
The one-hour procedure requires no handling or disposal of organic waste. RNA binds to a silica membrane and impurities are removed by washing through vacuum filtration or centrifugation. High-purity total RNA is eluted into RNase-free water; ideal for use in a variety of applications, including RT-PCR, qPCR, northern blotting, and nuclease protection assays.
Improved plate design
The PureLink Pro 96 total RNA Purification Kit provides optimized semi-skirted 96-well plates(Figure 3) for improved performance and results. In addition, the plate nozzles with annular collar prevent cross-contamination of samples and improve drying of the silica membrane to prevent ethanol carryover. In addition, sample processing can be performed manually using a benchtop vacuum manifold, by centrifugation (at 2250 x g), or with an automated liquid handling platform. The semi-skirted plate design is compatible with most vacuum manifolds on robotic workstations.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type RNA Wash Buffer I
Quantity 5 x 1 L