missing translation for 'onlineSavingsMsg'
Learn More

Veolia Water Systems™ Purification Cartridge

Product Code. 11970427
Click to view available options
For Use With (Equipment):
For PURELAB Classic Mk2
For PURELAB Option-Q
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11970427

Brand: Veolia Water Systems™ LC163

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Reliable, robust, easy-to-use general water purification cartridge and filter kits ideal for removing ionic contaminants to achieved specific water quality.

  • General water purification cartridge and filter kits
  • Deionization cartridge option Q
  • Purification cartridge
  • Removes ionic contaminants to achieve specified water quality

Specifications

For Use With (Application) Removes ionic contaminants to achieve specified water quality
For Use With (Equipment) For PURELAB Option-Q
Quantity 1 Each
Type Cartridge
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.