Learn More
Euro Protection™ Purofort™ Safety Boots
Product Code. 15839236
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
Each
Product Code. 15839236
Brand: Euro Protection™ 9PUSG46
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.
  • Flexible and lightweight for use all day long
  • Cold insulation
  • Tear and wear resistant
  • Purofort™ upper
  • Polyamide lining
  • Polyurethane insole
  • Steel toe cap
  • Steel, puncture-resistant sole
  • Purofort™ outsole
  • Suitable for use in farming industry
Specifications
Color Green
For Use With (Application) Agriculture/Farming
Liner Material Polyamide
Size 46
Sole Features Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Sole Material Purofort™
Toe Material Steel
Weight (Metric) 900 g per boot in size 9.5
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Boot