Learn More
Euro Protection™ Purofort™ Safety Boots
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
Each
Description
- Flexible and lightweight for use all day long
- Cold insulation
- Tear and wear resistant
- Purofort™ upper
- Polyamide lining
- Polyurethane insole
- Steel toe cap
- Steel, puncture-resistant sole
- Purofort™ outsole
- Suitable for use in farming industry
Specifications
Specifications
| Color | Green |
| For Use With (Application) | Agriculture/Farming |
| Liner Material | Polyamide |
| Size | 47 |
| Sole Features | Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole. |
| Sole Material | Purofort™ |
| Toe Material | Steel |
| Weight (Metric) | 900 g per boot in size 9.5 |
| Certifications/Compliance (Europe) | EN ISO 20345 : 2011 |
| Product Type | Safety Boot |