Learn More
Cole-Parmer™ Hand-Operated Vacuum Pump
Product Code. 11770009 Shop All Cole-Parmer Products
Click to view available options
Flow Rate:
16 mL/stroke
Unit Size:
Each
Product Code. 11770009
Brand: Cole-Parmer™ 7930111
Only null left
Add to basket
Add to basket
Produce up to 25 in. Hg vacuum or test for pressure leaks with this vacuum pump. Cole-Parmer™ Hand-Operated Vacuum Pumps are sealed and self-lubricated for a long, maintenance-free life.
  • Vacuum gauge is graduated in both centimeters and inches of mercury
  • Built-in trigger enables you to release vacuum without disconnecting the pump
  • PVC is lightweight and easy to handle
Specifications
Max. Vacuum 0.85 bar
Material PVC body, PTFE tap/Polycarbonate
Flow Rate 16 mL/stroke
Overall Height (Metric) 241 mm
Overall Length (Metric) 191 mm
Overall Width (Metric) 51 mm
Pump Type Vacuum Pump
Maximum Pressure 0.85 bar