Learn More
Dostmann Electronic™ PVC Precision Probes
Click to view available options
Sensor Types:
Class 1/10
Class 1/3
Unit Size:
Each
Description
- Temperature Sensor Pt100 6000-1074
- Pt100, Cl. 1/10 DIN -200...+450 °C 300 x 3,0 mm
- Immersion probe for measuring in liquid and powderd materials
- Sensor Class 1/10, Pt100(ceramic), 4-wire, DIN IEC 751,
- tube
- V2A or inconel, mineral-insulated, with handle, thermal stabilized
Specifications
Specifications
| Type | Probe, Precision |
| Material | PVC |
| Sensor Types | Class 1/10 |
| Accuracy | ± 0.03°C |
| Temperature Range (Metric) | -200°C to +450°C |
| Length (Metric) | 300 mm |