Learn More
Masterflex™ PVDF Barbed NPT Male Pipe Adapter Elbows
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30704-37
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11806743
  • 56.51€ / Pack of 10
Estimated Shipment
13-07-2021
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Translucent
Autoclavable
1 21/32 in.
1/2 in.
1/2 in.
Autoclave or Ethylene Oxide
1 23/32 in.
1/4 NPT(M) x 1/2 in. ID
PVDF
Barbed NPT Male Pipe Adapter Elbows
1/2 in.
1/4 in.
1/4 in.