Learn More
Cole-Parmer™ PVDF Manifold, 8 port
Product Code. 11745288
Click to view available options
Tubing Size:
1/2 in. x 3/8 in.
1/4 in. x 1/8 in.
Unit Size:
Each
Product Code. 11745288
Brand: Cole-Parmer™ 3152353
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Expand your flow and line capacity with these manifolds. Choose the number of ports and connections to suit your needs.
  • Constructed of durable PVDF.
  • Ideal for applications requiring excellent chemical compatibly.
  • Available in a variety of configurations
Specifications
Material PVDF
Overall Length (English) 8.75 in.
Overall Length (Metric) 22.23 cm
Min. Operating Temperature (English) -65°F
Min. Operating Temperature (Metric) -54°C
Tubing Size 1/2 in. x 3/8 in.
Inlet 1/2 in. NPT(F)
Outlet Port 3/8 in. NPT(F)
Width (Metric) 3.81 cm
Width (English) 1.5 in.
Height (Metric) 3.81 cm
Height (English) 1.5 in.
Maximum Pressure 110 psi
Max. Operating Temperature (English) 230°F
Max. Operating Temperature (Metric) 110°C
Product Type Manifold