Learn More
Cole-Parmer Cleanroom Panel-Mount Female Luer to Hose Barb Fittings, Straight Adapter, Nylon
Product Code. 30053487 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/32 in.
Color:
Natural
White
Unit Size:
Pack of 10
Product Code. 30053487
Brand: Cole-Parmer 5011569
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Material Nylon
Packaging Quantity 10/Pk.
Inlet Female Luer, Hose Barb
Color White
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.