Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Product Code. 11991518
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Height (Metric):
57 mm
68 mm
76 mm
85 mm
105 mm
130 mm
160 mm
210 mm
280 mm
315 mm
Diameter (Metric) Outer:
25 mm
30 mm
42 mm
51 mm
64 mm
85 mm
105 mm
131 mm
166 mm
187 mm
Closure Size:
15
22
30
42
Unit Size:
Each
Pack of 10
Pack of 20
Pack of 5
Pack of 50
Product Code. 11991518
Brand: Pyrex™ 1137/18M
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
  • PBT screw caps with PTFE faced sealing disc
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ Borosilicate Glass
  • Highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
TRUSTED_SUSTAINABILITY
Specifications
Closure Material SVL™ Phenolic, PTFE-faced with rubber sealing disc
Capacity (Metric) 3000 mL
Material Borosilicate Glass
Height (Metric) 315 mm
Includes Approximate graduations
Shape Conical
Graduated Yes
Closure Type Screw Cap
Diameter (Metric) Outer 187 mm
Closure Size 42