Learn More
Pyrex™ Borosilicate Glass Filter tube
Pyrex™ Borosilicate glass filter tubes are perfect for applications with Pyrex™ short stem funnels.
Brand: Pyrex 2110/08M
126.35 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.41000kg
Description
Pyrex™ Borosilicate glass filter tubes are perfect for applications with Pyrex™ short stem funnels.
- For small scale filtrations
- Ideal for use with Pyrex™ short stem funnels
Specifications
Tube Filter | |
6mm | |
Short-stemmed funnel Pyrex™ |
22mm | |
Pyrex Glass, Borosilicate Glass |