Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
6 L
10 L
20 L
Diameter (Metric) Outer:
50 mm
65 mm
85 mm
105 mm
131 mm
166 mm
207 mm
236 mm
279 mm
345 mm
Unit Size:
Each
Pack of 10
Description
- Complies with ISO 1773
- Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
Specifications
Specifications
| Certifications/Compliance | ISO 1773 |
| Capacity (Metric) | 6 L |
| Material | Borosilicate Glass |
| Height (Metric) | 355 mm |
| Bottom Shape | Round |
| Diameter (Metric) Outer | 236 mm |
| Closure Size | 53 |