Learn More
Corning™ PYREX™ Cloning Cylinders
Product Code. 11977595 Shop All Corning Products
Click to view available options
Diameter (Metric):
6 mm
8 mm
10 mm
Height (Metric):
8 mm
10 mm
Unit Size:
Pack of 125
Product Code. 11977595
Brand: Corning™ 31666
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Create identical cells by surrounding a single cell or group of cells
Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an ­isolated well.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Diameter (Metric) 6 mm
Height (Metric) 8 mm
Sterility Non-sterile
Diameter (English) 0.23 in.
Product Type Cloning Cylinder
Height (English) 0.31 in.