Learn More
Corning™ PYREX™ Disposable Glass Serological Pipets in Flip-Top Canister Pack, Sterile, Plugged
Shop All Corning Products
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Unit Size:
Pack of 400
Pack of 500
Description
These pipets are calibrated 'To Deliver' with blow-out. They are packed in a flip-top canister to keep the pipets racked. These pipet canisters come sealed in polyethylene bags for sterility. ISO color-coding is printed directly on the tear strip for easy identification by size. Reference: ASTM E-714.
Specifications
Specifications
| Aspirator Plug | Yes |
| Calibrated | Yes |
| Certifications/Compliance | ASTM E714 |
| Material | Borosilicate Glass |
| Sterility | Sterile |
| Graduated | Yes |
| Graduations | 0.01 mL |
| Length (English) | 10.7 in. |
| Negative Graduations | 0.2 mL |
| Tip Style | Round |
| Show More |