Learn More
Pyrex™ Rotaflo™ Stopcock for Aspirator
For use with Aspirator Bottles 1565: series with Rotaflo™ PTFE key.
Brand: Rotaflo GP6/10W
103.55 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.09000kg
Description
- Stopcock for use with Aspirator Bottles 1565: series with Rotaflo™ PTFE key
Specifications
10 mm | |
1 | |
Aspirator Bottles 1565 series | |
PTFE |