Learn More
Pyrex™ T-bore Plain arms 3-way Stopcock with PTFE Key
3-way Stopcock with PTFE Key
Brand:  Pyrex SST3025
159.60 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10775701
  • 168.00€ / Pack of 5
Estimated Shipment: 02-08-2024
to see stock.
Specifications
Plain Arms Stopcock
2.5 mm
14.5 mm
PTFE Key