missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,4-dicarboxylic acid, 98%, may cont. up to ca 10% water, Thermo Scientific™

Product Code. 11477027
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11477027

Brand: Thermo Scientific Alfa Aesar A14409.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an in vitro and in cell inhibitor, as well as a known inhibitor of the histone lysine demethylases. 2,4-Pyridinedicarboxylic acid has been used in a study to determine that ruthenium(II) complexes exert antimetastatic effects on several tumor cell lines in vitro, achieved mostly by the effect on cell adhesion, migration and angiogenesis. It blocks the activity of 2-OG oxygenases, which include certain lysine demethylases and a variety of hydroxylases (e.g., prolyl, collagen, lysyl). 2,3 2,4-PDCA inhibits several Jumonji domain-containing lysine demethylases when used at low micromolar concentrations. Through its effects on hydroxylases, including prolyl hydroxlase 1 (IC50 = 1.5 µM), It modulates hypoxia inducing factor turnover, collagen synthesis, and plant cell wall formation. It can inhibit zinc-dependent enzymes, like metallo-β-lactamase.8 2,4-PDCA also affects and is translocated by organic anion transporters.

Solubility
Soluble in water at 20°C 4.5 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-80-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00149413,MFCD00006296
InChI Key MJIVRKPEXXHNJT-UHFFFAOYSA-N
Synonym 2,4-pyridinedicarboxylic acid, lutidinic acid, 2,4-lutidinic acid, 2,4-pyridinedicarboxylicacid, hydrate 1:1, 4nrq, pd2, 4-carboxypicolinic acid, pubchem16314, pyridine carboxylate, 6a, acmc-1bnb9
PubChem CID 10365
ChEBI CHEBI:44737
IUPAC Name pyridine-2,4-dicarboxylic acid
SMILES OC(=O)C1=CC=NC(=C1)C(O)=O

Specifications

Melting Point ∼245°C (decomposition)
pH 2.5 to 3.5
Quantity 5 g
Beilstein 131631
Solubility Information Soluble in water at 20°C 4.5g/L.
Formula Weight 167.12
Percent Purity 98%
Assay may cont. up to ≈10% water
Chemical Name or Material Pyridine-2,4-dicarboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.