missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium dichromate, 98%

Código de producto. 10348050 Tienda Thermo Scientific Chemicals Productos
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Tamaño de la unidad:
1kg
250g
50g
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10348050

Marca: Thermo Scientific Acros 200882500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 20039-37-6 | C10H12Cr2N2O7 | 376.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 20039-37-6
Molecular Formula C10H12Cr2N2O7
Molecular Weight (g/mol) 376.2
InChI Key LMYWWPCAXXPJFF-UHFFFAOYSA-P
Synonym pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
PubChem CID 2724130
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
SMILES C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Especificaciones

Melting Point 152.0°C to 153.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Quantity 250 g
Fieser 09,399; 10,335; 12,419; 14,269; 15,57
Merck Index 15,8087
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmf, dmso and dimethylacetamide -fairly, soluble in acetonenitrile - sparingly soluble in, methylene chloride, chloroform and ether - pract., insol.in hexane,toluene,ethyl ether,ethyl acetate
Formula Weight 376.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium dichromate
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Pyridinium dichromate
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H350i-May cause cancer by inhalation.
  • H272-May intensify fire; oxidiser.
  • H228-Flammable solid.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado