missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pyronin Y, Certified
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 250.0°C to 260.0°C |
Absorption Ratio | 1.23 to 1.7 |
Dye Content | 45% min. |
Beilstein | 18, V,10, 181 |
Merck Index | 15, 8119 |
Solubility Information | Solubility in water: soluble. |
Infrared Spectrum | Authentic |
Lambda Max | 546 to 551nm |
Formula Weight | 302.79 |
Grade | Pure |
Show More |
Safety and Handling
Product Identifier
- Pyronin Y
- Warning
- Germ cell mutagenicity Category 2
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'