missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15407547
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
7126-39-8
C5H5NO
95.10
MFCD03426733
CHNYVNOFAWYUEG-UHFFFAOYSA-N
pyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxaldehyde, pyrrole-3-carbaldehyde, 3-formylpyrrole, 3-formyl-1h-pyrrole, 3-formyl-pyrrole, pyrrol-3-aldehyde, pyrrole-3-aldehyde, pubchem18051, acmc-209oht
640308
1H-pyrrole-3-carbaldehyde
O=CC1=CNC=C1
This item is not returnable. View return policy
7126-39-8
C5H5NO
95.10
MFCD03426733
CHNYVNOFAWYUEG-UHFFFAOYSA-N
pyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxaldehyde, pyrrole-3-carbaldehyde, 3-formylpyrrole, 3-formyl-1h-pyrrole, 3-formyl-pyrrole, pyrrol-3-aldehyde, pyrrole-3-aldehyde, pubchem18051, acmc-209oht
640308
1H-pyrrole-3-carbaldehyde
O=CC1=CNC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 65°C to 68°C
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 95.1
Percent Purity 97%
Chemical Name or Material Pyrrole-3-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'