Learn More
Izon qEV2 Column
Product Code. 15948090
Encompass_Preferred
Click to view available options
:
For use with EV/exsosome isolation
Product Code. 15948090
Brand: Izon SP4
This item requires a price update prior to being ordered.
Request a Price for this item here.
qEV provides the most rapid and effective EV/exsosome isolation system available. qEV is based on Size Exclusion Chromatography (SEC), and takes around 15 minutes to get a pure sample of intact exosomes. The resulting samples are consistent, standardisable and repeatable.
  • The qEV2 is designed for separating exosomes from a 2mL biological sample and comes with Luer Lock fittings. They will be of particular interest if you are working with larger fluid volumes. The Luer Lock fittings allow for many different configurations
Specifications
Description Izon qEV2
Column Type Size Exclusion Column
For Use With (Application) EV/exsosome isolation