missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Glass Bottle Beakers with Green Thermoset F217 and PTFE Caps

Product Code. p-4599683 Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
240 mL
Quantity:
24/Cs.
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 11800110

Brand: Qorpak™ 3460140

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Can be used for mixing, measuring and storage

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Specifications

Capacity 8 oz., 240 mL
Capacity (Metric) 240 mL
Capacity (English) 8 oz.
Color Clear
Diameter (English) 2.44 in.
Material Glass
Quantity 24/Cs.
Closure Color Green
Height (Metric) 127 mm
Height (English) 5 in.
Closure Liner F217, PTFE
Closure Material Thermoset
Closure Size 58-400 mm
Diameter (Metric) 62 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.