Learn More
Hellma™ QS Quartz Suprasil High Precision Macro Absorption Cells With PTFE Stopper
Product Code. 10475031
Click to view available options
Capacity (Metric):
350 μL
7000 μL
Unit Size:
Each
Pack of 2
Product Code. 10475031
Brand: Hellma™ 110140
Only null left
Specifications
Type Macro Absorption Cell
Capacity (Metric) 350 μL
Material Quartz Suprasil
Closure Material PTFE
Width (Metric) Interior 9.5mm
Path Length 1 mm
Wavelength Range 200 to 2500 nm
Height (Metric) 52 mm
Width (Metric) 12.5 mm
For Use With (Application) Photometric and Colorimetric Measurements