Learn More
Macherey Nagel Qualitative Analysis Test Papers
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 70 mm
Quantity:
200 Strips
Unit Size:
Pack of 200
Description
- Test papers allow the qualitative determination of ions and chemical compounds
- They are used to detect whether compounds tested for are present above the specific detection limit
- Some of the papers have specific applications
Specifications
Specifications
| For Use With (Application) | Silver ions (AG +) |
| Quantity | 200 Strips |
| Type | Qualitative analysis test strips |
| Dimensions (L x W) | 20 x 70 mm |