Learn More
Socorex™ Qualitix™ Tipfill Refill System
Intended for easy rack loading. Socorex™ Qualitix™ Tipfill Refill System are filled trays for instant transfer into a rack without risk of contamination.
Brand:  Socorex™ 3070.0200T
Additional Details : Weight : 0.78000kg
 View more versions of this product
Product Code. 15760649
  • 62.30€ / Pack of 960
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Easy rack loading.
  • Instant transfer without contamination risk.
  • Smart, re-closable Tipfill box reduces storage space, waste volume, and shipping costs.
Specifications
Specifications
200 μL
Natural
96/Rack
Single-use
200μL
Beveled
10/PK, 10PK/CS
Qualitix