missing translation for 'onlineSavingsMsg'
Learn More

Witeg™ Quartz Glass Tall-Form Beakers

Offers excellent transparency and high temperature reliability. Witeg™ Quartz Glass Low-Form Beakers are available in a range of capacities.

Brand:  Witeg™ 8601025

Additional Details : Poids : 1.00000kg

 View more versions of this product

Product Code. 13487547

  • 37.95€ / Each


Expédition estimée
23-10-2020
Add to basket

Description

Description

  • Tall form beakers with spout
  • Transparent quartz silica glass contruction DIN12332
  • Not graduated
  • Range of volumes
  • High resistance to thermal shock
  • Good chemical resistance
Specifications

Specifications

Yes
30mm
Quartz Silica Glass
Beaker
Tall form
25 mL
55mm
Videos
Documents