Learn More
Hellma™ Quartz Suprasil 4 Faces Cuveet
Click to view available options
Wavelength Range:
200 to 2500 nm
Unit Size:
Pack of 2
Specifications
Specifications
| Type | Cuvette |
| Description | 4 Faces |
| Material | Quartz Suprasil |
| Quantity | 2Pack |
| Wavelength Range | 200 to 2500 nm |